missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Afamin Antibody (120), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90199AF700
This item is not returnable.
View return policy
Description
Afamin Monoclonal specifically detects Afamin in Human samples. It is validated for ELISA.
Specifications
| Afamin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 173 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 120 | |
| ELISA | |
| afamin, ALB2alpha-Alb, ALBAalpha-albumin, ALF, Alpha-Alb, Alpha-albumin, MGC125338, MGC125339 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Afamin (P43652; Met1-Asn599). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction