missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Afadin Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149408
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.
Specifications
| Afadin | |
| Polyclonal | |
| Unconjugated | |
| Afdn | |
| 5033403D15Rik; Af6; Af-6; Afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; afadin; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; FLJ34371; Gm314; I-afadin; MLL-AF6; MLLT4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; protein Af-6; RP3-431P23.3; RP3-470B24.4; S-afadin | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 4301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P55196 | |
| Afdn | |
| E. coli-derived recombinant human Afadin/AF-6, Asn217-Pro392. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction