missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Afadin Monoclonal Antibody (851204)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561875

Product Code. 30534447

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Afadin
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P55196
Afdn
E. coli-derived recombinant human Afadin/AF-6, Asn217-Pro392.
100 μg
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
851204
Unconjugated
Afdn
5033403D15Rik; Af6; Af-6; Afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; afadin; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; FLJ34371; Gm314; I-afadin; MLL-AF6; MLLT4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; protein Af-6; RP3-431P23.3; RP3-470B24.4; S-afadin
Mouse
Protein A/G
RUO
4301
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.