missing translation for 'onlineSavingsMsg'
Learn More

Afadin/AF-6 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18402594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402594 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402594 Supplier Novus Biologicals Supplier No. NBP122996DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Afadin/AF-6 Polyclonal antibody specifically detects Afadin/AF-6 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Afadin/AF-6
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AF64, afadin, FLJ34371, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1475 and 1525 of human AF6 (myeloid/lymphoid or mixed-lineage leukemia translocated to, 4 ) using the numbering given in entry BAA32484.1 (GeneID 4301).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4301
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.