missing translation for 'onlineSavingsMsg'
Learn More

AF64394, MedChemExpress

Product Code. 16491487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491487 50 mg 50mg
16487997 5 mg 5mg
16445946 100 mg 100mg
16425288 10 mg 10mg
16403326 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16491487 Supplier Medchem Express Supplier No. HY12664
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AF64394 is a GPR3 inverse agonist, with a pIC50 of 7.3.

AF64394 is a GPR3 inverse agonist, with a pIC50 of 7.3.

Specifications

Chemical Name or Material AF64394
CAS 1637300-25-4
Color White
Purity Grade Notes Research
Molecular Formula C21H20ClN5O
Quantity 50 mg
Solubility Information DMSO : ≥ 125 mg/mL (317.36 mM)
SMILES ClC1=CC(OC(C)C)=C(CNC2=CC(C3=CC=CC=C3)=NC4=NC=NN42)C=C1
Molecular Weight (g/mol) 393.87
Formula Weight 393.87
Percent Purity 98.02%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.