missing translation for 'onlineSavingsMsg'
Learn More

AF4 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18428606 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428606 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428606 Supplier Novus Biologicals Supplier No. NBP128728AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AF4 Polyclonal antibody specifically detects AF4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AF4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AF-4, AF4/FMR2 family member 1, AF4/FMR2 family, member 1, AF4myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 2, ALL1-fused gene from chromosome 4 protein, FEL, MLLT2MGC134969, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 2, PBM1, pre-B-cell monocytic leukemia partner 1, Protein AF-4, Protein FEL, Proto-oncogene AF4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human proto-oncogene AF4 using the numbering given in entry NP_005926.1 (GeneID 4299).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4299
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.