missing translation for 'onlineSavingsMsg'
Learn More

AF38469, MedChemExpress

Product Code. 16485358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485358 10 mg 10mg
16458077 5 mg 5mg
16445986 100 mg 100mg
16401557 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485358 Supplier Medchem Express Supplier No. HY12802

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AF38469 is a selective, orally bioavailable Sortilin inhibitor with an IC50 value of 330 nM.

AF38469 is a selective, orally bioavailable Sortilin inhibitor with an IC50 value of 330 nM.

Specifications

Chemical Name or Material AF38469
CAS 1531634-31-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H11F3N2O3
Quantity 10 mg
Solubility Information DMSO : ≥ 43 mg/mL (132.61 mM)
SMILES O=C(NC1=CC=CC(C)=N1)C2=CC=C(C(F)(F)F)C=C2C(O)=O
Molecular Weight (g/mol) 324.25
Formula Weight 324.25
Percent Purity 98.05%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.