missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF10 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-49867
This item is not returnable.
View return policy
Description
AF10 Polyclonal specifically detects AF10 in Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| AF10 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| AF10MGC75086, ALL1-fused gene from chromosome 10 protein, DKFZp686E10210, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 10, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 10, protein AF-10, type I AF10 protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8028 | |
| Rat | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_004632.1 | |
| MLLT10 | |
| Peptide with sequence C-RLEDTTARFTNAN corresponding to internal region according to NP_001009569.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_004632.1; NP_001009569.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction