missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AF-DX 116

Product Code. 18722644
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18722644

Brand: Tocris Bioscience™ 1105/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective M2 antagonist

Selective M2 muscarinic receptor antagonist. Ki values are 64, 417, 786, 211 and 5130 nM for human recombinant M2, M1, M3, M4 and M5 muscarinic receptors, respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102394-31-0
Molecular Formula C24H31N5O2

Specifications

Quantity 10 mg
Formula Weight 421.54
Purity >98%
Molecular Weight (g/mol) 421.545
Chemical Name or Material AF-DX 116
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.