Learn More
Lovibond™ AF 415 Alkalinity Caustic/P MINIKIT
Product Code. 10441541
Click to view available options
Range:
20 to 500 mg/L as CaCO3
Unit Size:
Each
Product Code. 10441541
Brand: Lovibond™ 414150
AF 415 Alkalinity Caustic/P MINIKIT measures P alkalinity 20-500 mg/L as CaCO3 includes reagents for approximately 30 tests
They are user-friendly and suitable for a wide variety of applications in water analysis.Thanks to various test procedures, the MINIKITs are one thing above all else: a reliable companion.
Various test procedures
For the MINIKITs various test procedures are available:
Choose between manual test procedures, Turbidity or Tablet Count Method or the so-called Speed test.
Intuitive application
Whether professional or layman: with the MINIKIT everyone achieves error-free and reliable results.
Long durability
The reagents used in the MINIKITs have a shelf life of 5 years. And best of all - they are not subject to any transport restrictions.
Directly usable
In addition to the test kit in a sturdy plastic box, the delivery content also includes a measuring beaker, the necessary accessories and tablet reagents for approx. 30 tests.
Specifications
Analytes P Alkalinity
Range 20 to 500 mg/L as CaCO3
Type Test Kit