missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Aerotap Tube for ASP-620 Series Aspirator Bottle
Description
- 12mm bore tap
- Air-vented
Specifications
Specifications
| Plug Bore | 12 mm |
| Description | Carboy tap |
| Fits Tubing ID | 12 mm |
| For Use With (Equipment) | ASP-620 Series Aspirator Bottle |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?