missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Aerosolv™ 360 System
Description
Includes
Premium puncturing unit, high-efficiency filter, wire, and goggles.
Requires
55 Gallon Drum
- Enclosed system—dual seal chamber eliminates the potential for content spray back, prevents VOC release and reduces gunk build up
- High-efficiency combination coalescing carbon filter captures odors, and prevents potentially harmful VOCs from being released into the air—extended filter life reduces change outs to save time and money
- Alignment cone and two-way pressure controlled, non-sparking vent pin offers the most efficient point of puncture—greatly reduces cleanup and minimizes maintenance
- Intuitive, ergonomic activation wheel design is easy to use, and with no lever handle extending over the side of the drum, removes the chance for snagged clothing or tipped drums
- Ideal for high volume applications - safely converts spent aerosol cans from solid hazardous waste to a non-hazardous, recyclable material.
- Threading directly into a standard 2-in (51-mm) bung on a 30- (110-liter) or 55-gallon (200-liter) drum, the housing accommodates a wide variety of the most common sized cans, including domed mini, standard, and jumbo.
- Sleek design with no protruding lever handle—mechanical activation wheel with rack and pinion gear increases torque making can processing easy and fast.
- Integrated lock out system prevents unauthorized use and tamper-proof retention plate secures the unit to the drum.
- Non-sparking puncture pin pierces the can and directs the liquid flow straight into the drum for a quick, controlled evacuation.
- Non-stick coating on housing minimizes static transfer to the unit while at the same time reduces clean up.
- Attach the two-stage high-efficiency combination filter to the ¾-in (19-mm) bung to capture VOCs from any remaining propellants.
- Replaceable, proprietary polymer coalescer in filter base captures the residual liquid in the gas before it reaches the activated carbon, helping extend the life of the filter.
- The top activated carbon filter cartridge features adsorption indicator which reacts to the adsorption of VOCs to indicate when the filter cartridge is ready for replacement.
- Built-in counter tracks system use for maintenance.
- Includes safety goggles and an anti-static ground wire with alligator clip.
- One 55-gallon (200-liter) drum holds approximately 4,000 spent aerosol cans.
- Punctured, discharged, cans do not meet the definition of hazardous waste per EPA 40 CFR 261.23(a)(6), and thus can be treated as recyclable scrap metal.
Compliance
EPA Compliant
Warranty
1 year
Specifications
Specifications
| Certifications/Compliance | EPA compliant |
| Color | Black, Green |
| Quantity | 1/Pk. |
| Autoclavable | Not Autoclavable |
| Includes | Premium puncturing unit, high-efficiency filter, wire, and goggles. |
| For Use With (Application) | Recycling aerosol cans |
| For Use With (Equipment) | 55 gallon drum |
| Height (English) | 17 in. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?