missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Aeromonas Aminopeptidase Recombinant Protein, PeproTech®
Description
100-10-1MG will be provided as 2 x 500 μg (100-10-500UG) Recombinant Aeromonas Aminopeptidase is a 31.4 kDa protein containing 291 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The enzyme (29 KD) aeromonas aminopeptidase is used in the processing of pharmaceutical proteins produced by genetic engineering as well as for physical and structural investigations and for sequence and amino-terminal determinations. This exopeptidase recognizes a specific stop sign at -X- Pro and requires a free a-amino group in the L-configuration. It is therefore suitable for the removal of the redundant N-terminal methionine often added to engineered proteins.
Specifications
Specifications
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Molecular Weight (g/mol) | 31.4 kDa |
| Name | Aeromonas Aminopeptidase |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Common Name | Aeromonas Aminopeptidase |
| Biological Activity | Sequentially cleaves N-terminal amino acids except E, D, and X-P. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?