missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Aeromonas Aminopeptidase Recombinant Protein, PeproTech®

Product Code. 17810793
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
500 μg
Unit Size:
100µg
1mg
500µg
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17810793 500 μg 500µg
17850143 100 μg 100µg
17880783 2 x 500 μg 1mg
30088079 2 x 500 μg Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17810793 Supplier Gibco™ Supplier No. 10010500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

100-10-1MG will be provided as 2 x 500 μg (100-10-500UG) Recombinant Aeromonas Aminopeptidase is a 31.4 kDa protein containing 291 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The enzyme (29 KD) aeromonas aminopeptidase is used in the processing of pharmaceutical proteins produced by genetic engineering as well as for physical and structural investigations and for sequence and amino-terminal determinations. This exopeptidase recognizes a specific stop sign at -X- Pro and requires a free a-amino group in the L-configuration. It is therefore suitable for the removal of the redundant N-terminal methionine often added to engineered proteins.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation protein with no preservative
Molecular Weight (g/mol) 31.4 kDa
Name Aeromonas Aminopeptidase
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name Aeromonas Aminopeptidase
Biological Activity Sequentially cleaves N-terminal amino acids except E, D, and X-P.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MPPITQQATV TAWLPQVDAS QITGTISSLE SFTNRFYTTT SGAQASDWIA SEWQALSASL PNASVKQVSH SGYNQKSVVM TITGSEAPDE WIVIGGHLDS TIGSHTNEQS VAPGADDDASGIAAVTEVIR VLSENNFQPK RSIAFMAYAA EEVGLRGSQD LANQYKSEGK NVVSALQLDM TNYKGSAQDV VFITDYTDSN FTQYLTQLMD EYLPSLTYGF DTCGYACSDH ASWHNAGYPAAMPFESKFND YNPRIHTTQD TLANSDPTGS HAKKFTQLGL AYAIEMGSAT G
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.