missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HALYARD™ AERO CHROME™ High-Performance Surgical Gown
High-Performance Surgical Gown combines comfort, softness and breathability with high level protection against pathogens and fluids, including chemotherapy drugs.
£176.40 - £229.24
Specifications
| Product Type | Surgical Gown |
|---|---|
| No. of Pockets | None |
| Material | Cool Shield Copolymer |
| Color | Silver |
| Closure Type | Hook and Loop Neck with Waist Ties |
| Product Code | Brand | Size | Sterility | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Size | Sterility | Price | Quantity & Availability | ||||
|
30274682
|
HALYARD™
48392 |
Large | Non-sterile |
£199.01
Pack of 40 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274725
|
HALYARD™
48393 |
X-Large | Non-sterile |
£176.40
Pack of 32 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274736
|
HALYARD™
48394 |
2X-Large | Non-sterile |
£181.95
Pack of 32 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274651
|
HALYARD™
48395 |
Large | Sterile |
£202.56
Pack of 30 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274705
|
HALYARD™
48396 |
X-Large | Sterile |
£203.86
Pack of 28 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274656
|
HALYARD™
48397 |
Large, X-Long | Sterile |
£213.22
Pack of 30 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274668
|
HALYARD™
48398 |
X-Large, X-Long | Sterile |
£229.24
Pack of 30 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274657
|
HALYARD™
48399 |
Large, X-Long | Non-sterile |
£192.21
Pack of 36 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274722
|
HALYARD™
48400 |
X-Large, X-Long | Non-sterile |
£187.50
Pack of 32 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274767
|
HALYARD™
48401 |
2X-Large, X-Long | Non-sterile |
£181.24
Pack of 30 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274704
|
HALYARD™
48402 |
2X-Large, X-Long | Sterile |
£229.10
Pack of 28 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
|
30274695
|
HALYARD™
48403 |
2X-Large | Sterile |
£218.91
Pack of 28 |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||
Description
- Culmination of HALYARD™'s expertise in designing high-performance garments for critical applications in Healthcare, the AERO CHROME™ gowns offer an exceptional combination of Protection, Breathability, and Comfort.
- Red collar: Easily identify a high-performance gown
- Breathable back panel: maximizes airflow for cool comfort
- Soft, light, and breathable fabric: keeps staff protected yet cool and comfortable
- Low linting: helps reduce the risk of wound contamination and infection
- Meets the High Performance per EN13795-1:2019: ensures superior protection
- Meets AAMI Level 4 standard: provides the highest level of fluid and microbial protection in the critical zones
- No breakthrough detected after 240 minutes of exposure to 52 chemotherapy drugs
- Raglan Sleeves for extra comfort and free movements
- Available Sterile and Non Sterile
- Sterile gown packed individually and with 2 towels for added convenience
- Non Sterile gowns packed bulk in a bag to reduce packaging
Specifications
| Surgical Gown | |
| Cool Shield Copolymer | |
| Hook and Loop Neck with Waist Ties | |
| Knit | |
| Raglan Sleeve | |
| Not Autoclavable |
| None | |
| Silver | |
| Single-use | |
| Yes | |
| Regulation (EU) 2017/745 Class I, EN 13795, AAMI level 4, ASTM D6978-05 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title