missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEG-1/MTDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13860-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody got two bands about 70 kDa and 80 kDa in western blotting, possibly due to posttranslational modification. This antibody is specific to AEG-1/MTDH.
Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.Specifications
| AEG-1/MTDH | |
| Polyclonal | |
| Unconjugated | |
| MTDH | |
| 3D3, 3D3/LYRIC, AEG 1, AEG1, LYRIC, LYRIC/3D3, metadherin, Metastasis adhesion protein, MTDH, Protein LYRIC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170910, 67154, 92140 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80WJ7, Q86UE4, Q9Z1W6 | |
| Mtdh | |
| AEG-1/MTDH-Specific Fusion Protein Ag4840 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction