missing translation for 'onlineSavingsMsg'
Learn More

AEC, 50X, TRC

Product Code. 30330579
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330579 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330579 Supplier Toronto Research Chemicals Supplier No. TRCA7717231G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Amino-N-ethylcarbazole
CAS 132-32-1
Molecular Formula C14 H14 N2
Quantity 1 g
Synonym 9-Ethyl-9H-carbazol-3-amine, 3-Amino-9-ethylcarbazole, 3-Amino-N-ethylcarbazole, 9-Ethyl-3-carbazolamine, 9-Ethyl-9H-carbazol-3-ylamine, 9-Ethyl-9H-carbazole-3-amine, AEC, AEC (chromogen), NSC 67709, NSC 96640, AEC, 50X
InChI Formula InChI=1S/C14H14N2/c1-2-16-13-6-4-3-5-11(13)12-9-10(15)7-8-14(12)16/h3-9H,2,15H2,1H3
SMILES CCn1c2ccccc2c3cc(N)ccc13
IUPAC Name 9-ethylcarbazol-3-amine
Molecular Weight (g/mol) 210.27
Formula Weight 210.1157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.