missing translation for 'onlineSavingsMsg'
Learn More

AEBSF Hydrochloride (White to Off-white Crystals), Fisher BioReagents

Product Code. 10193483
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Packaging:
Poly Micro Tube
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193483 500 mg Poly Micro Tube 500mg
10081553 50 mg Poly Micro Tube 50mg
10563165 100 mg Poly Micro Tube 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193483 Supplier Fisher BioReagents Supplier No. BP635500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This specific, irreversible inhibitor of serine proteases is a stable, nontoxic alternative to PMSF.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H11ClFNO2S
Molecular Weight (g/mol) 239.689
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl

Specifications

Melting Point 175°C
Color White
Assay Percent Range ≥98.0 %
Packaging Poly Micro Tube
Quantity 500 mg
Formula Weight 239.7
Percent Purity ≥98 (HPLC)
Physical Form Powder or Solid
Chemical Name or Material AEBSF Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.