missing translation for 'onlineSavingsMsg'
Learn More

AEBSF Hydrochloride, TRC

Product Code. 30327817
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327817 Supplier Toronto Research Chemicals Supplier No. TRCA32530050MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material AEBSF Hydrochloride
CAS 30827-99-7
Molecular Formula C8H11ClFNO2S
Quantity 50 mg
Synonym 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride, p-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride, Pefabloc, Pefabloc SC, Pefabloc SC Plus
InChI Formula InChI=1S/C8H10FNO2S.ClH/c9-13(11,12)8-3-1-7(2-4-8)5-6-10;/h1-4H,5-6,10H2;1H
SMILES c1c[n+]2c(cc1)c3[n+](cccc3)CC2
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
Molecular Weight (g/mol) 239.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.