missing translation for 'onlineSavingsMsg'
Learn More

AEBSF hydrochloride, MedChemExpress

Product Code. 16465996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465996 100 mg 100mg
16431056 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465996 Supplier Medchem Express Supplier No. HY12821
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AEBSF hydrochloride is an irreversible inhibitor of serine proteases, such as chymotrypsin, kallikrein, plasmin, thrombin, and trypsin.

AEBSF hydrochloride is an irreversible inhibitor of serine proteases, such as chymotrypsin, kallikrein, plasmin, thrombin, and trypsin.

Specifications

Chemical Name or Material AEBSF hydrochloride
Physical Form Solid
CAS 30827-99-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H11ClFNO2S
Solubility Information DMSO : 125 mg/mL (521.51 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (417.21 mM)
SMILES O=S(C1=CC=C(CCN)C=C1)(F)=O.[H]Cl
Molecular Weight (g/mol) 239.69
Formula Weight 239.69
Percent Purity 95.78%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.