missing translation for 'onlineSavingsMsg'
Learn More

Melford AEBSF HCl

Product Code. 30017957
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
50mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017957 5 g 5g
30017956 1 g 1g
30017955 500 mg 500mg
30017953 50 mg 50mg
30017954 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30017957 Supplier Melford Supplier No. A200105.0
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specific and irreversible inhibitor of serine proteases.

Specific and irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF.

Specifications

CAS 30827-99-7
Molecular Formula C8H10FNO2S-HCl
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.