missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AEBP2 Monoclonal Antibody (2D7B6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531612
This item is not returnable.
View return policy
Description
MA5-31612 has been tested in indirect ELISA.
AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates 'Lys-9' and 'Lys-27' residues of histone H3.
Specifications
| AEBP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q6ZN18 | |
| AEBP2 | |
| Purified recombinant fragment of human AEBP2 (AA: 358-495) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 2D7B6 | |
| Unconjugated | |
| AEBP2 | |
| adipocyte enhancer-binding protein 2; Adipocyte enhancer-binding protein 2 homolog; AE binding protein 2; AE(adipocyte enhancer)-binding protein 2; AE-binding protein 2; AE-binding protein 2 homolog; aebp2; AU023766; B230313N05Rik; LOW QUALITY PROTEIN: zinc finger protein AEBP2; si:dkey-158p11.2; wu:fk42f11; wu:fk52h12; zgc:63755; Zinc finger protein AEBP2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 121536 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction