missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBP2 Antibody (3E3C10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | AEBP2 |
|---|---|
| Clone | 3E3C10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18209671
|
Novus Biologicals
NBP2-61835 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605688
|
Novus Biologicals
NBP2-61835-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AEBP2 Monoclonal specifically detects AEBP2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| AEBP2 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Adipocyte enhancer-binding protein 2, AE binding protein 2, AE(adipocyte enhancer)-binding protein 2, AE-binding protein 2, MGC17922, zinc finger protein AEBP2 | |
| AEBP2 | |
| IgG1 | |
| Protein G purified |
| 3E3C10 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| Unconjugated | |
| Mouse | |
| 121536 | |
| Purified recombinant fragment of human AEBP2 (AA: 358-495) expressed in E. Coli. | |
| Primary | |
| 54.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title