missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBP2 Antibody (2D7B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61822
This item is not returnable.
View return policy
Description
AEBP2 Monoclonal specifically detects AEBP2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| AEBP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| Adipocyte enhancer-binding protein 2, AE binding protein 2, AE(adipocyte enhancer)-binding protein 2, AE-binding protein 2, MGC17922, zinc finger protein AEBP2 | |
| Mouse | |
| 54.5 kDa | |
| 0.1 mL | |
| 121536 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2D7B6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AEBP2 | |
| Purified recombinant fragment of human AEBP2 (AA: 358-495) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction