Learn More
AE2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26332-1-AP-150UL
This item is not returnable.
View return policy
Description
The AE2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human AE2. This antibody recognizes human antigen. The AE2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| AE2 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 6522 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 533 μg/mL | |
| Rabbit | |
| Primary | |
| Human |