missing translation for 'onlineSavingsMsg'
Learn More

ADX88178, MedChemExpress

Product Code. 16477558
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477558

Brand: Medchem Express HY18654

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ADX88178 is a potent metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM) with an EC50 of 4 nM for human mGluR4.

ADX88178 is a potent metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM) with an EC50 of 4 nM for human mGluR4.

Specifications

Chemical Name or Material ADX88178
Physical Form Solid
CAS 1235318-89-4
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C12H12N6S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 16.67 mg/mL (61.21 mM; Need ultrasonic)
SMILES CC1=C(C2=CNN=C2)N=C(NC3=NC=CC(C)=N3)S1
Molecular Weight (g/mol) 272.33
Formula Weight 272.33
Percent Purity 99.6%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.