missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ADX 10059 hydrochloride

Product Code. 18055432
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18055432

Brand: Tocris Bioscience™ 4416/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Negative allosteric modulator at mGlu5

ADX 10059 hydrochloride is reported to be a negative allosteric modulator at the mGlu5 receptor. Analog of MPEP (Cat. No. 1212).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 757949-98-7
Molecular Formula C15H14ClFN2

Specifications

Quantity 50 mg
Formula Weight 276.74
Purity >99%
Molecular Weight (g/mol) 276.739
Chemical Name or Material ADX 10059 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.