Learn More
Advillin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20956-1-AP-150UL
 View more versions of this product
Product Code. 16880535
  • £349.00 / 150µL
Description
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Specifications
Specifications
Advillin
Polyclonal
Unconjugated
AVIL
ADVIL, advillin, AVIL, DOC6
Rabbit
Antigen Affinity Chromatography
RUO
10677, 11567
-20°C
Liquid
Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75366, O88398
AVIL
Advillin Fusion Protein Ag15114
150 μL
Primary
Human, Mouse