missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Advillin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20956-1-AP-20UL
This item is not returnable.
View return policy
Description
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.Specifications
| Advillin | |
| Polyclonal | |
| Unconjugated | |
| AVIL | |
| ADVIL, advillin, AVIL, DOC6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10677, 11567 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75366, O88398 | |
| AVIL | |
| Advillin Fusion Protein Ag15114 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction