missing translation for 'onlineSavingsMsg'
Learn More

Advanced Instruments Protinol™ Serum Osmometer Controls

Product Code. 15148616
Encompass
Change view
Click to view available options
For Use With (Equipment):
3320, 3300 Micro-osmometer
Unit Size:
Pack of 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15148616 3320, 3300 Micro-osmometer Pack of 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15148616 Supplier Advanced Instruments Supplier No. 3MA028
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quality control check for the clinical laboratory for osmometers from any manufacturer

  • Developed from bovine serum albumin — adjusted with nonprotein material
  • Produce consistent, reliable results in the human serum range
  • Run controls daily to check and document the osmometer's performance
  • Set of three values: 240 ±5mOsm/kg H2O (abnormal low), 280 ±5mOsm/kg H2O (normal), and 320 ±5mOsm/kg H2O (abnormal high)

Specifications

Certifications/Compliance NIST, ISO 9000, CLIA, CAP
Content And Storage 2°C to 8°C
Concentration 240, 280, 320 mOsm/kg H2O
For Use With (Equipment) 3320, 3300 Micro-osmometer
Sample Type Serum
Type 3-Level Serum Control Kit
Includes 3mL 3 vials per level
Intended Use Use in evaluating performance of osmometer in clinical laboratories testing of Human Serum samples
Quantity 9 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.