Learn More
Specac™ Advanced High Temperature High Pressure Cell for FT-IR
Click to view available options
Includes:
Reflectance mode wedge pressurised window assembly
Description
This product permits analysis of solid samples in transmission, specular reflectance and decomposition modes; and process gases in static or flow transmission mode. Sample temperatures of up to 800°C can be achieved and the cell can operate at pressures from vacuum to 1000 psi. Also includes reflectance mode wedge pressurised window assembly. Compatible with all FTIR instruments.
Specifications
Specifications
| Description | Compatible with all FTIR instruments. |
| Includes | Reflectance mode wedge pressurised window assembly |