missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADU-S100 ammonium salt, MedChemExpress
Description
ADU-S100 ammonium salt (MIW815 ammonium salt), an activator of stimulator of interferon genes (STING), leads to potent and systemic tumor regression and immunity.
Specifications
Specifications
| Chemical Name or Material | ADU-S100 ammonium salt |
| CAS | 1638750-96-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H30N12O10P2S2 |
| Quantity | 10 mg |
| Synonym | MIW815 ammonium saltML RR-S2 CDA ammonium salt |
| Solubility Information | DMSO : 15 mg/mL (20.70 mM; Need ultrasonic and warming) ∣H2O : 0.5 mg/mL (0.69 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | OC1([H])[C@](O[P@](S)(OC[C@](O[C@@H](N2C3=NC=NC(N)=C3N=C2)[C@@H]4O)([H])[C@@]4([H])O5)=O)([H])[C@H](N6C7=NC=NC(N)=C7N=C6)O[C@]1([H])CO[P@]5(S)=O.N.N |
| Molecular Weight (g/mol) | 724.6 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?