missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADSSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106947
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
ADSSL1 is a muscle isozyme of adenylosuccinate synthase (EC 6.3.4.4), which catalyzes the initial reaction in the conversion of inosine monophosphate (IMP) to adenosine monophosphate (AMP).
Specifications
| ADSSL1 | |
| Polyclonal | |
| Unconjugated | |
| ADSSL1 | |
| adenylosuccinate synthase like 1; adenylosuccinate synthetase 1, muscle; adenylosuccinate synthetase isozyme 1; Adenylosuccinate synthetase like 1; adenylosuccinate synthetase, basic isozyme; Adenylosuccinate synthetase, muscle isozyme; Adenylosuccinate synthetase-like 1; Adss; adSS 1; Adss1; Adssl1; AI528595; AMPSase 1; IMP--aspartate ligase 1; M-type adenylosuccinate synthetase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 122622 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q8N142 | |
| ADSSL1 | |
| Synthetic peptide directed towards the middle region of human ADSSL1 (aa 144-194). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction