missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADSS Recombinant Rabbit Monoclonal Antibody (K01_1M45)

Product Code. 30828459
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828459 100 μL
30825915 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828459 Supplier Invitrogen™ Supplier No. MA563266

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ADSS Recombinant Monoclonal Antibody for ICC/IF, Western Blot

This gene encodes the enzyme adenylosuccinate synthetase which catalyzes the first committed step in the conversion of inosine monophosphate to adenosine monophosphate. A pseudogene of this gene is found on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADSS
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1M45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ADSS2
Gene Accession No. P30520, P46664
Gene Alias ADEH; adenylosuccinate synthase; adenylosuccinate synthetase (Ade(-)H-complementing); adenylosuccinate synthetase 2, non muscle; adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; adenylosuccinate synthetase, liver isozyme; adenylosuccinate synthetase, non muscle; adenylosuccinate synthetase, non-muscle isozyme; Adss; ADSS 2; Adss2; AI314886; AMPSase 2; AS; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase
Gene Symbols ADSS2
Host Species Rabbit
Immunogen Recombinant protein of human AdSS 2.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11566, 159
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.