missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADSS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112351
This item is not returnable.
View return policy
Description
ADSS Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes the enzyme adenylosuccinate synthetase which catalyzes the first committed step in the conversion of inosine monophosphate to adenosine monophosphate. A pseudogene of this gene is found on chromosome 17.
Specifications
| ADSS | |
| Polyclonal | |
| Unconjugated | |
| ADSS2 | |
| ADEH; adenylosuccinate synthase; adenylosuccinate synthetase (Ade(-)H-complementing); adenylosuccinate synthetase 2, non muscle; adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; adenylosuccinate synthetase, liver isozyme; adenylosuccinate synthetase, non muscle; adenylosuccinate synthetase, non-muscle isozyme; Adss; ADSS 2; Adss2; AI314886; AMPSase 2; AS; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11566, 159 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30520, P46664 | |
| ADSS2 | |
| Recombinant Human Adenylosuccinate synthetase isozyme 2 protein (1-280AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction