missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADSL Recombinant Rabbit Monoclonal Antibody (8J9E10)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557909

 View more versions of this product

Product Code. 30284636

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAAGGDHGSP DSYRSPLASR YASPEMCFVF SDRYKFRTWR QLWLWLAEAE QTLGLPITDE QIQEMKSNLE NIDFKMAAEE EKRLRHDVMA HVHTFGHCCP KAAGIIHLGA TSCYVGDNTD LIILRNALDL LLPKLARVIS RLADFAKERA SLPTLGFTHF QPAQLTTVGK RCCLWIQDLC MDLQNLKRVR DDLRFRGVKG TTGTQASFLQ LFEGDDHKVE QLDKMVTEKA GFKRAFIITG QTYTRKVDIE VLSVLASLGA SVHKICTDIR LLANLKEMEE PFEKQQIGSS AMPYKRNPMR SERCCSLARH LMTLVMDPLQ TASVQWFERT LDDSANRRIC LAEAFLTADT ILNTLQNISE GLVVYPKVIE RRIRQELPFM ATENIIMAMV KAGGSRQDCH EKIRVLSQQA ASVVKQEGGD NDLIERIQVD AYFSPIHSQL DHLLDPSSFT GRASQQVQRF LEEEVYPLLK PYESVMKVKA ELCL.

ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ADSL
Recombinant Monoclonal
1.66 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P30566
Adsl
Recombinant protein (or fragment).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
8J9E10
Unconjugated
Adsl
2510006M18Rik; Adenylosuccinase; adenylosuccinate lyase; adenylosuccinate lyase 1; Adl; Adsl; AMPS; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; ASase; ASL; null; test; unnamed protein product
Rabbit
Protein A
RUO
158
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.