Learn More
Invitrogen™ ADSL Recombinant Rabbit Monoclonal Antibody (8J9E10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557910
Description
Immunogen sequence: MAAGGDHGSP DSYRSPLASR YASPEMCFVF SDRYKFRTWR QLWLWLAEAE QTLGLPITDE QIQEMKSNLE NIDFKMAAEE EKRLRHDVMA HVHTFGHCCP KAAGIIHLGA TSCYVGDNTD LIILRNALDL LLPKLARVIS RLADFAKERA SLPTLGFTHF QPAQLTTVGK RCCLWIQDLC MDLQNLKRVR DDLRFRGVKG TTGTQASFLQ LFEGDDHKVE QLDKMVTEKA GFKRAFIITG QTYTRKVDIE VLSVLASLGA SVHKICTDIR LLANLKEMEE PFEKQQIGSS AMPYKRNPMR SERCCSLARH LMTLVMDPLQ TASVQWFERT LDDSANRRIC LAEAFLTADT ILNTLQNISE GLVVYPKVIE RRIRQELPFM ATENIIMAMV KAGGSRQDCH EKIRVLSQQA ASVVKQEGGD NDLIERIQVD AYFSPIHSQL DHLLDPSSFT GRASQQVQRF LEEEVYPLLK PYESVMKVKA ELCL.
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.
Specifications
| ADSL | |
| Recombinant Monoclonal | |
| 1.66 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P30566 | |
| Adsl | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8J9E10 | |
| Unconjugated | |
| Adsl | |
| 2510006M18Rik; Adenylosuccinase; adenylosuccinate lyase; adenylosuccinate lyase 1; Adl; Adsl; AMPS; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; ASase; ASL; null; test; unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 158 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.