missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADSL Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£218.00 - £436.00
Specifications
| Antigen | ADSL |
|---|---|
| Clone | 001 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17729823
|
Invitrogen™
MA543474 |
50 μL |
£218.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17789603
![]() |
Invitrogen™
MA543054 |
100 μL |
£436.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human ADSL/Adenylosuccinate Lyase
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.Specifications
| ADSL | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| 2510006M18Rik; Adenylosuccinase; adenylosuccinate lyase; adenylosuccinate lyase 1; Adl; Adsl; AMPS; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; ASase; ASL; null; test; unnamed protein product | |
| Adsl | |
| IgG | |
| Protein A | |
| Antibody |
| 001 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P30566 | |
| 158 | |
| Recombinant Human ADSL/Adenylosuccinate Lyase protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Adsl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title