missing translation for 'onlineSavingsMsg'
Learn More

ADRP/Perilipin 2 Mouse anti-Human, Clone: 1G5D4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60340-1-IG-20UL

 View more versions of this product

Product Code. 16807455

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Anti-ADRP antibody is a reliable and sensitive marker for lipid droplets. Enhanced expression of ADRP is linked to diseases with abnormal lipid storage, including hepatic steatosis, atherosclerosis and diabetes. Immunohistochemistry of ADRP may facilitate

Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.
Specifications

Specifications

ADRP/Perilipin 2
Monoclonal
1.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99541
PLIN2
ADRP/Perilipin 2 Fusion Protein Ag7643
20 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
1G5D4
Unconjugated
PLIN2
ADFP, Adipophilin, ADRP, Perilipin 2, PLIN2
Mouse
Protein G
RUO
123
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.