missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adropin Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA522995

Product Code. 13268258

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Suggested positive control: Adropin Overexpression lysate, mouse brain tissue lysate. 88% sequence identity with bovine protein.

Adropin is involved in the regulation of glucose homestasis and lipid metabolism. It is a secreted protein encoded by the Energy Homeostasis Associated gene (ENHO) found in the brain and liver. In the liver, the protein has been found to increase in response to high-fat foods and decrease after fasting. The protein seems to play a role in controlling the activity of other metabolic genes, specifically those involved in the production of lipids from carbohydrates.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adropin
Polyclonal
Unconjugated
ENHO
1110065P19Rik; 2310040A07Rik; Adropin; adropin-like; C9orf165; energy homeostasis associated; energy homeostasis-associated protein; ENHO; GAAI470; LOC100912292; RGD1565232; UNQ470; UNQ470/PRO830
Rabbit
Antigen affinity chromatography
RUO
375704, 69638
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1.11 mg/mL
PBS with 30% glycerol and 0.1% sodium azide
Q6UWT2, Q8K1D8
ENHO
Synthetic peptide made to an internal portion of human Adropin (within residues 10-60).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.