missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Adropin Polyclonal antibody specifically detects Adropin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Adropin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470 |
| Host Species | Rabbit |
| Immunogen | ADROPIN antibody was raised against an 18 amino acid peptide near the carboxy terminus of human ADROPIN. The immunogen is located within the last 50 amino acids of ADROPIN. Amino Acid Squence: PEKAPPPQKKPSHEGSYL |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?