missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adropin Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26387AF350
This item is not returnable.
View return policy
Description
Adropin Polyclonal specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Adropin | |
| Polyclonal | |
| Western Blot | |
| adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470 | |
| Rabbit | |
| 0.1 mL | |
| 375704 | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ENHO | |
| Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-ProtNo. Q6UWT2] | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction