missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADRM1 Recombinant Rabbit Monoclonal Antibody (24GB4700)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556955
This item is not returnable.
View return policy
Description
ADRM1 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| ADRM1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ADRM1 | |
| 110 kDa cell membrane glycoprotein; 1110063P18Rik; 2510006J17Rik; AA408205; adhesion regulating molecule 1; adhesion-regulating molecule 1; ADRM1; Arm1; ARM-1; AU043535; cell membrane glycoprotein, 110000M(r) (surface antigen); glycoprotein 110; GP110; hRpn13; M(r) 110,000 surface antigen; MGC29536; proteasomal ubiquitin receptor ADRM1; Proteasome regulatory particle non-ATPase 13; Rpn13; rpn13 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11047 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB4700 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16186 | |
| ADRM1 | |
| A synthesized peptide derived from human ADRM1 (aa 123-136). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction