Learn More
Invitrogen™ ADRM1 Recombinant Rabbit Monoclonal Antibody (24GB4700)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556956
 View more versions of this product
Product Code. 30276628
  • £126.00 / 20µL
Description
ADRM1 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ADRM1
Recombinant Monoclonal
Unconjugated
ADRM1
110 kDa cell membrane glycoprotein; 1110063P18Rik; 2510006J17Rik; AA408205; adhesion regulating molecule 1; adhesion-regulating molecule 1; ADRM1; Arm1; ARM-1; AU043535; cell membrane glycoprotein, 110000M(r) (surface antigen); glycoprotein 110; GP110; hRpn13; M(r) 110,000 surface antigen; MGC29536; proteasomal ubiquitin receptor ADRM1; Proteasome regulatory particle non-ATPase 13; Rpn13; rpn13 homolog
Rabbit
Affinity chromatography
RUO
11047
-20°C
Liquid
Flow Cytometry, Western Blot
24GB4700
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q16186
ADRM1
A synthesized peptide derived from human ADRM1 (aa 123-136).
20 μL