missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRM1 Antibody (34C2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59402R
This item is not returnable.
View return policy
Description
ADRM1 Monoclonal antibody specifically detects ADRM1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| ADRM1 | |
| Monoclonal | |
| DyLight 550 | |
| 000 surface antigen, 110 kDa cell membrane glycoprotein, adhesion regulating molecule 1, Adhesion-regulating molecule 1, ARM1, ARM-1, Gp110, GP110hRpn13, M(r) 110, MGC29536, proteasomal ubiquitin receptor ADRM1, Proteasome regulatory particle non-ATPase 13, regulatory particle non-ATPase 13, Rpn13, Rpn13 homolog | |
| Recombinant human ADRM1 (1-407aa) purified from E. coli | |
| 0.1 mL | |
| Cancer | |
| 11047 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 34C2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction