missing translation for 'onlineSavingsMsg'
Learn More

Adriforant hydrochloride, MedChemExpress

Product Code. 16497666
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497666

Brand: Medchem Express HY19705B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist binding affinity (Ki=2.4 nM) and is also a functional (Ki=1.56 nM) antagonist.

Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist binding affinity (Ki=2.4 nM) and is also a functional (Ki=1.56 nM) antagonist.

Specifications

Chemical Name or Material Adriforant hydrochloride
Physical Form Solid
CAS 2096455-90-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H25Cl3N6
For Use With (Application) COVID-19-immunoregulation
Synonym PF-3893787 hydrochloride
Solubility Information H2O : 100 mg/mL (269.01 mM; Need ultrasonic) ∣DMSO : ≥ 83.33 mg/mL (224.16 mM)
SMILES NC1=NC(N2C[C@H](NC)CC2)=CC(NCC3CC3)=N1.[H]Cl.[H]Cl.[H]Cl
Molecular Weight (g/mol) 371.74
Formula Weight 371.74
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.