missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adrenomedullin R/ADMR/GPR182 Antibody (528563) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10293JF525
This item is not returnable.
View return policy
Description
Adrenomedullin R/ADMR/GPR182 Monoclonal antibody specifically detects Adrenomedullin R/ADMR/GPR182 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
Specifications
| Adrenomedullin R/ADMR/GPR182 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| ADMR, adrenomedullin receptor, AMR, AM-R, G protein-coupled receptor 182,7TMR, G10D, gamrh, G-protein coupled receptor 182, hrhAMR, MGC34399 | |
| HEK293 human embryonic kidney cell line transfected with human Adrenomedullin R/ADMR, Accession # O15218 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 528563 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 11318 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction