Learn More
Adrenocorticotrophic Hormone (ACTH) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Adrenocorticotrophic Hormone (ACTH) Ab-1, Mouse Monoclonal Antibody.
£96.16 - £416.00
Specifications
| Antigen | Adrenocorticotrophic Hormone (ACTH) Ab-1 |
|---|---|
| Clone | AH26 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12671467
|
Epredia
MS-452-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12681467
|
Epredia
MS-452-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12601477
|
Epredia
MS-452-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12691467
|
Epredia
MS-452-P1 |
500 μL | Concentrated | 200μg/mL | |||||
|
12661467
|
Epredia
MS-452-P |
1 mL | Concentrated | 200μg/mL | |||||
|
12631477
|
Epredia
MS-452-R7 |
7 mL | Prediluted | ||||||
Description
Host Species: Mouse
Clone: AH26
Isotype: IgG1
Species Reactivity: Human. It shows a broad species reactivity.
Epitope: aa 1-24
Immunogen: A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH
Positive Control: Normal pituitary gland or pituitary adenoma
Cellular Localization: Cytoplasmic Ab-1 is excellent for staining of routine formalin-fixed, paraffin-embedded tissues. It labels corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas. NeoMarkers' Ab-1 and Ab-2 may provide a pair for ELISA
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Adrenocorticotrophic Hormone (ACTH) Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG1 |
| AH26 | |
| Monoclonal | |
| Mouse | |
| Steroids Hormones and Growth Factors | |
| A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.