missing translation for 'onlineSavingsMsg'
Learn More

Adrenaline Tartrate, Mikromol™

Product Code. 18081560
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081560 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18081560 Supplier Mikromol Supplier No. MM0614.00
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Adrenaline Tartrate
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Adrenaline tartrate
CAS 51-42-3
Analyte or Component Names Adrenaline tartrate
Shipping Condition Room Temperature
Molecular Formula C9 H13 N O3 . C4 H6 O6
Certifications/Compliance ISO 17025
Synonym Adrenaline Tartrate, L-Adrenaline Tartrate, 4-[(1R)-1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt), (R)-4-[1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Adrenaline tartrate (1:1), Tartaric acid compd. with adrenaline, (-)-Adrenaline acid tartrate, (-)-Adrenaline bitartrate, (-)-Adrenaline hydrogen tartrate, (-)-Adrenaline tartrate, (-)-Epinephrine (+)-bitartrate, (-)-Epinephrine bitartrate, Adrenalin bitartrate, Adrenaline acid tartrate, Adrenaline bitartrate, Adrenaline hydrogen tartrate, Asmatane Mist, Asthmahaler Epitrate, Bronitin Mist, Bronkaid Mist Suspension, Epinephrine bitartrate, Epinephrine d-bitartrate, Epinephrine hydrogen tartrate, Epinephrine tartrate, Epitrate, L-Adrenaline bitartrate, Medihaler-Epi, Primatene Mist Suspension, R-(-)-Epinephrine bitartrate, l-Adrenaline bitartrate, l-Adrenaline d-bitartrate, l-Adrenaline hydrogen tartrate, l-Adrenaline tartrate, l-Epinephrine bitartrate, l-Epinephrine d-bitartrate, (-)-3,4-Dihydroxy-alpha-[(methylamino)methyl]benzyl alcohol (+)-tartrate (1:1) (salt)
InChI Formula InChI=1S/C9H13NO3.C4H6O6/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;5-1(3(7)8)2(6)4(9)10/h2-4,9-13H,5H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t9-;1-,2-/m01/s1
SMILES CNC[C@H](O)c1ccc(O)c(O)c1.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Molecular Weight (g/mol) 333.29
Quantity 500 mg
Formula Weight 333.106 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.