missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ADR LQ Transport Sign
Black and white, diamond-shaped Limited Quantity transport signs.
Brand: Brady™ 813992
Additional Details : Weight : 0.75200kg
Description
- Replaces LQ for all transport except plane
Specifications
Not Autoclavable | |
Black on White | |
Laminated Polyester | |
Facility Identification, Hazard Communication Identification | |
Diamond | |
0.099 mm | |
100 mm |
Permanent Acrylic | |
Gloss | |
Non-sterile | |
250 Labels | |
-40°C to +110°C | |
100 mm |